Learn More
Cole-Parmer Hose Barb Fitting, Tee Union, HDPE, Cleanroom Packed, 3/8" ID; 10/Pk
Supplier:  Cole-Parmer 5011110
Encompass_Preferred
Use these high-quality PVC-free fittings in biomedical, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Easily combine or split fluid flow using tee unions. A single barb on each end of the fitting creates a tight seal to keep the tubing in place. These high-density polyethylene (HDPE) fittings are softer than nylon, polypropylene, and Kynar. They exhibit excellent resistance to chemicals but have lower thermal resistance than other materials.
Catalog No. 50-255-8118