Learn More
StatLab Hot Foil Tape for TBS/Thermo Microwriter (1.125" x 400') 6 rolls per case
SDP
Supplier:  StatLab SLHFT
Encompass_Preferred
Thermal transfer ribbon, used for automated cassette printing. Prints up to 20,000 cassettes per roll. Designed for use on TBS SHUR/Mark cassette printers and Thermo MicroWriter cassette printers.
Catalog No. NC1588321
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.