Learn More
West Chester™ Heavy Weight Cotton Hot Mill Gloves with Burlap Liner
Catalog No. 19821557 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Large
Catalog No.
19-821-557 Large
1 options
This item is not returnable. View return policy
Catalog No. 19-821-557 Supplier West Chester™ Supplier No. 7900BL
This item is not returnable. View return policy
Multi layer cotton canvas construction offers comfort, insulation, and superior heat protection. Used in forging, hot metal handling, castings, molds and other operations that require heat protection.
  • Burlap lining for increased heat protection
  • 100% cotton outer shell
  • Clute cut provides a roomy fit
  • Straight thumb increases comfort in closed-fisted work
  • Nap-out finish for heat resistance
  • Knuckle strap for extra protection
  • Gauntlet cuff for added forearm protection and fast removal
  • Washable for longer wear
Specifications
Exterior Finish Nap-out