Learn More
Hach Company Hot Plate Stirrer, 7x7 in., digital, 115 Vac
Supplier: Hach Company 2881600
Easy-to-read digital temperature settings features a flashing warning (CAUTION! HOT TOP) when the surface exceeds 50ºC. Stirring range of 60-1200 RPM.Low-profile design to improve safety. Reflective, white ceramic top is smooth, seamless and resistant to alkali/acid solutions. Detachable cord. Built-in support rod holder (order separately). Operating temperature range: 5 to 540°C (41 to 1004°F).