Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-Biosciences Hot Start Taq DNA Polymerase 2X Master Mix
SCI_ED

Catalog No. S03152
Change view
Click to view available options
No. of Reactions:
100
500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
S03152 500
S03153 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S03152 Supplier G-Biosciences Supplier No. 7861965
Add to Cart
Add to Cart

Hot start Taq DNA Polymerase 2X Master Mix is used for routine PCR, qPCR, genotyping and high GC content template DNA amplification with GC enhancer.

  • G-biosciences Hot start Taq DNA Polymerase 2x master mix is ready to use premix which contains hot start Taq DNA Polymerase, dNTPs and optimized buffer with stabilizers.
  • Hot start Taq DNA Polymerase is chemically modified that possesses a 5' - 3' polymerase activity and a 5'-flap endonuclease activity.
  • Hot start Taq DNA Polymerase leads to complete inactivation of the polymerase until the initial heat activation step at the start of PCR.
  • Hot start Taq DNA Polymerase reduces non-specific amplification during PCR setup and increases PCR specificity and sensitivity.

Specifications

Detection Method Gel Electrophoresis
Form Liquid
GC-Rich PCR Performance High
PCR Method Standard PCR, Direct PCR, qPCR, Multiplex qPCR, 1-step RT-qPCR
Reaction Speed Fast
Label or Dye Biotin, 6-FAM, ROX, SYBR Green, TAMRA
Product Type DNA Polymerase Master Mix
For Use With (Application) Hot-start PCR, Multiplex PCR, High-fidelity PCR, Site-directed Mutagenesis, Next-Generation Sequencing (NGS)
No. of Reactions 500
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.