Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-Biosciences Hot Start Taq DNA Polymerase
SCI_ED

Catalog No. p-200080565
Change view
Click to view available options
No. of Reactions:
200
1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
S03154 1000
S03155 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S03154 Supplier G-Biosciences Supplier No. 7861963
Only null left
Add to Cart
Add to Cart

Hot start Taq DNA Polymerase is used for routine PCR, qPCR, genotyping and high GC content template DNA amplification with GC enhancer.

  • G-biosciences hot start Taq DNA Polymerase is chemically modified that possesses a 5' - 3' polymerase activity and a 5'-flap endonuclease activity.
  • Hot start Taq DNA Polymerase reduces non-specific amplification during PCR setup and increases PCR specificity and sensitivity.

Specifications

Detection Method qPCR
Form Liquid
GC-Rich PCR Performance High
PCR Method Standard PCR, Direct PCR, qPCR, Multiplex qPCR, 1-step RT-qPCR
Reaction Speed Standard
Label or Dye Biotin, 6-FAM, ROX, SYBR Green, TAMRA
Product Type DNA Polymerase
For Use With (Application) Hot-start PCR, Multiplex PCR, Genotyping, DNA Sequencing, Next-Generation Sequencing (NGS)
No. of Reactions 1000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.