Learn More
Analytical Sales & Services HOTSLEEVE SMART COLUMN HEATER
Supplier:  Analytical Sales & Services HSI1508L
Encompass_Preferred
HotSleeve+ Self-Calibrating "Smart" Column Heater, Includes: 1.5" ID x 8" Heater Only. Temperature Range: 5ºC Above Ambient to 85ºC.
Catalog No. NC2380963
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.