Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ HotStart-IT Binding Protein

Catalog No. 71194400UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
400 μg (200 x 25 μL rxns)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-194-400UG 400 μg (200 x 25 μL rxns)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71-194-400UG Supplier Thermo Scientific™ Supplier No. 71194400UG
Add to Cart
Add to Cart

HotStart-IT Binding Protein is the active component in a novel hot start technology called primer sequestration.

HotStart-IT Binding Protein is the active component in the novel hot start technology known as primer sequestration. In primer sequestration, hot start PCR methods reduce or eliminate non-specific primer-extension products formed at lower temperatures during assembly of PCR reactions. At these less stringent annealing temperatures, primers may bind non-specifically, which often leads to unwanted amplification products and primer-dimers. To resolve this problem, we have produced a high-quality DNA binding protein that is especially useful at sequestering primers at lower temperatures, making them unavailable for use by a polymerase (see figure).

Features

  • Room temperature reaction setup
  • Minimizes amplification of non-specific products and primer-dimers (see figure)
  • Ideal for complex templates and multiplex reactions
  • No extensive heating step required
  • Free from detectable non-specific nucleases
  • Technology is portable to a polymerase of choice

This primer-sequestration technique effectively blocks DNA synthesis from mis-priming events at lower temperatures. Following the initial denaturation step during PCR, the protein is inactivated, and the primers are free to participate in the amplification reaction.

HotStart-IT Binding Protein performs well in many standard PCR reaction buffers and has been designed for PCR applications that demand high specificity and sensitivity. PCR with Taq DNA Polymerase and HotStart-IT Binding Protein shifts production of primer-dimers to a specific target of 306 bp from 1 ng of human genomic DNA relative to Taq DNA Polymerase alone.

Application

  • Hot-Start PCR amplification

Notes

  • One microgram of HotStart-IT Binding Protein sequesters about 5 pmol of primers.
  • HotStart-IT Binding Protein is sourced from recombinant protein expressed in E. coli.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Hot-start PCR
Content And Storage • HotStart-IT Binding Protein, 400 μg

Store at -20°C.
Shipping Condition Dry Ice
Compatible Buffer 1X PCR Reaction Buffer
Quantity 400 μg (200 x 25 μL rxns)
Product Type HotStart-IT Binding Protein

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.