Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ HotStart™ Storage Reaction Tubes

Catalog No. 2123661 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Line:
HotStart Micro 50 Strips
Volume (Metric):
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Line Volume (Metric)
21-236-61 HotStart Micro 50 Strips 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-236-61 Supplier Thermo Scientific™ Supplier No. 6210

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Eliminate tedious oil or wax overlays with the thin-walled Thermo Scientific™ HotStart™ Storage Reaction Tubes.

Features:
  • Bead is stable at room temperature
  • Assures synchronous reaction startup
  • PCR product can remain in tube for later analysis
  • Hinged caps provide snug seal
  • Presterilized; RNase- and DNase-free
Recommended for: Complex DNA or cDNA, very low copy number targets, more than 30 thermal cycles, multiple PCR or RT-PCR
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 0.2 mL
For Use With (Equipment) Thermal Cyclers
Product Line HotStart Micro 50 Strips
Sterility Sterile
Closure Type Flat Cap
Reaction Volume 25 to 50 μL
Product Type Strips
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.