Learn More
Hourglass International™ HCE™ 3-Ply Cleanroom Face Masks with Ties
Cleanroom face masks are manufactured for long-wearing comfort and protect Class 100/ISO 5 environments from human contaminants.
Supplier:  Hourglass International™ FM3EMEA23218CM
 View more versions of this product
Catalog No. 19-423-055
This item is not returnable. View return policy
Description
Description
  • Made from the cleanest 3-ply polyethylene and polypropylene materials
  • High bacterial and particle filtration efficiency
  • Four white SBPP ties
  • Soft, flexible nose piece
  • Ultra-low lint level design for critical environments
  • Manufactured and double-bagged in a cleanroom environment
  • Suitable for ISO 5 / Clean Class 100 and higher
Specifications
Specifications
Cleanroom Face Mask
Ties
Standard
Pleated
3-ply
>99%
Yes
18 cm
50/Bag, 2000/Cs.
HCE™
White
Polyethylene, Polypropylene
Non-sterile
Class 100 (ISO 5)
Single-use
Semiconductor, Disk Drive, Medical Device, Pharmaceutical, Biomedical
7 in.
FM3EMEA232-18CM
Double-bagged, Cleanroom Packaged
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.