Learn More
Honeywell™ Howard Leight™ Clarity™ C3 Ear Muffs
Catalog No. 19161268
Change view
Click to view available options
C3; Headband
Catalog No.
19-161-268 C3; Headband
1 options
Catalog No. 19-161-268 Supplier Honeywell™ Supplier No. 1011146
Improve worker safety by blocking noise, while voice and signal frequencies can be heard more naturally
  • Sound management technology enhances on-the-job communication through uniform attenuation
  • Noise Reduction Rating: 27dB
  • Blocks noise while voice, signal, alarm and other high frequencies can be heard more naturally, with less distortion, combating overprotection and worker isolation
  • Robust non-deforming dielectric construction withstands use and abuse, while protecting in electrical environments
  • Height adjustment remains fixed during wear
  • Snap-in ear cushions make replacement quick and easy
  • Non-deforming outer headband provides better comfort, minimizes pressure on head, withstanding rough treatment in the toughest workplaces
  • Inner-ventilated headband provides better positioning, minimizes pressure on the head
TRUSTED_SUSTAINABILITY
Specifications
Color Metallic Blue
Certifications/Compliance NRR 27
For Use With (Application) Agriculture and farming, building construction, forestry, general contracting, heavy construction, landscaping, lumber or wood products, metal fabricationPetrochemicalSteelMiningTransportationUtility/Waste Management
Noise Reduction Rating (NRR) 27 dB
Product Type Ear Muffs
Dielectric Yes
Material Plastic
Style Over-the-head