Learn More
Honeywell™ Howard Leight™ Hygiene Kit for QM24+™ Ear Muff
Catalog No. 19051331 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Cushion Color:
Black
Catalog No. Cushion Color
19-051-331 Black
1 options
Catalog No. 19-051-331 Supplier Honeywell™ Supplier No. HK4
  • $598.00
    $370.65 / Case of 20
    Save $227.35
    38% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Hygiene kit extends the performance and lifetime of the QM24+™ ear muff.
  • Includes one pair of replacement ear cushions and one pair of foam inserts
  • Extends performance and lifetime of ear muff
  • Recommended replacement every six months with general use, more frequently with heavy use
  • Snap-in ear cushions are standard on all Howard Leight ear muffs
Specifications
Cushion Material Foam, Vinyl
Cushion Color Black
For Use With (Equipment) Howard Leight™ QM24+™ Ear Muffs
Model HK4
Packaging Type Poly bag
Product Type Hygiene Kit
Material Foam