Learn More
Honeywell™ Howard Leight™ Source 400 Dispenser Ear Plug Refill
Catalog No. 19085490 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Laser Lite Refill
Catalog No.
19-085-490 Laser Lite Refill
1 options
Catalog No. 19-085-490 Supplier Honeywell™ Supplier No. LLLS4REFILL
Add to Cart
Add to Cart
Zip top refill bags are packaged in packs of 200 pairs. Replenish Howard Leight™ LS-400 Ear Plug Dispensers as a cost-effective solution while eliminating waste.
  • Convenient and environmentally-friendly refill bags for Howard Leight™ LS-400 ear plug dispenser
  • Choose from Laser Lite™ Max™ or MultiMax™ disposable foam ear plugs
  • Packaging includes 200 pairs per refill bag and 10 refill bags per case
Specifications
ANSI S3.1974 Yes
For Use With (Application) Agriculture, Chemical, Construction, Fire Protection, Forestry, Government, Law Enforcement, Manufacturing, MedicalMilitary, Mining, Municipal Services, Oil and Gas, Pharmaceutical, Steel and Metals, Transportation, Utilities, Welding
Noise Reduction Rating (NRR) 25/32 dB
Corded Uncorded
Plug Shape T-Shape
Product Type Laser Lite™ Refill for LS-400
Disposable Single-use