Learn More
Honeywell™ Howard Leight™ Matrix Ear Plugs
Catalog No. 19161065
Change view
Click to view available options
Uncorded; Orange; 29 db
Catalog No.
19-161-065 Uncorded; Orange; 29 db
1 options
Catalog No. 19-161-065 Supplier Honeywell™ Supplier No. MTX1OR
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
Matrix ear plugs provide uniform attenuation that blocks harmful noise while voice frequencies can be heard more naturally, combating worker isolation and overprotection on the job.
  • Patented no-roll design makes insertion fast and easy
  • Smooth outer skin and reduced diameter provide long-term comfort
  • Delivers instant protection upon proper insertion – no need to wait for foam to expand
  • Uniform attenuation profile blocks out noise while voice frequencies can be heard more naturally
TRUSTED_SUSTAINABILITY
Specifications
ANSI S3.1974 Yes
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Dual-density foam is easy to insert with a simple push no rolling required
High Visibility No
Packaging Type Poly bag
Pairs per Package 200 Pairs/Pk.
Plug Material Foam
Plug Color Orange
Noise Reduction Rating (NRR) 29 dB
Corded Uncorded
Plug Shape Cylinder
Disposable Single-use
Metal Detectable No