Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Howard Leight™ Matrix™ Single-Use Earplugs

Catalog No. 19911410
Change view
Click to view available options
Matrix Single-Use Earplugs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-911-410 Matrix Single-Use Earplugs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-911-410 Supplier Honeywell™ Supplier No. MTX30OR

No-roll design and enhanced communication

  • Smooth outer skin and reduced diameter provide long-term comfort
  • Delivers instant protection upon proper insertion — no need to wait for foam to expand
  • Uniform attenuation profile blocks out noise while voice frequencies can be heard more naturally
  • Three attenuation levels for targeted attenuation (Orange NRR 29, Green NRR 27 and Blue NRR 23)
  • Bulk refills for Leight Source 400 and Leight Source 500 Earplug Dispensers save time, waste and space
  • Color: Orange

TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Certifications/Compliance ANSI S3.19-1974
Size Universal
For Use With (Application) Smooth outer skin and reduced diameter provide long term comfort
Noise Reduction Rating (NRR) 29 dB
Corded Corded
Plug Shape Cylinder
Product Type Howard Leight™ Matrix Earplug
Disposable Single-use
Material TPE, Foam
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.