Learn More
Honeywell™ Howard Leight™ Quiet Band HYG Hearing Protectors
Catalog No. p-91386
Change view
Click to view available options
Model:
QB2HYG
QB3HYG
For Use With (Application):
Semi-Aural Protection for People Who Are Exposed to Moderate or Intermittent Noise
Supra-Aural Protection for People Who Are Exposed to Intermittent Noise
Noise Reduction Rating (NRR):
23 dB
25 dB
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model For Use With (Application) Noise Reduction Rating (NRR)
19-033-675 QB2HYG Supra-Aural Protection for People Who Are Exposed to Intermittent Noise 25 dB
19-033-408 QB3HYG Semi-Aural Protection for People Who Are Exposed to Moderate or Intermittent Noise 23 dB
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-033-675 Supplier Honeywell™ Supplier No. QB2HYG
Lightweight and portable hearing protectors with patented band design prevents ear pods from touching dirty or contaminated surfaces when set down. Choose a specific style QB1HYG, QB2HYG or QB3HYG to balance worker's desired comfort and protection.
  • Suspended pod design prevents contact with unsanitary surfaces
  • Pads made of soil-resistant, high-attenuating foam that can be easily cleaned and reused
  • Extra-wide ergonomic bands, packaged with an extra pair of pods
  • QB1HYG inner-aural protection fits inside opening of the ear canal (NRR 27 dB)
  • QB2HYG supra-aural protection fits partially inside opening of the ear canal (NRR 25 dB)
  • QB3HYG semi-aural protection fits outside the opening of the ear canal (NRR 23 dB)
TRUSTED_SUSTAINABILITY
Specifications
ANSI S3.1974 Yes
CSA Z94.2 Class BL
Size Universal
Headband Material Polypropylene
For Use With (Application) Supra-Aural Protection for People Who Are Exposed to Intermittent Noise
Model QB2HYG
Pairs per Package 1 Ea.
Plug Material Foam
Plug Color Orange
Noise Reduction Rating (NRR) 25 dB
Plug Shape Bell
Metal Detectable No
Headband Color Orange