Learn More
Honeywell™ Thunder T3 Ear Muff Hygiene Kit
Catalog No. 19161069
Change view
Click to view available options
:
T3 Hygiene Kit
Catalog No.
19-161-069 T3 Hygiene Kit
1 options
Catalog No. 19-161-069
Supplier: Honeywell™ 1010976
Extends performance and lifetime of the Thunder T3 Ear Muff. Includes one pair of snap-in ear cushions and one pair of foam ear cup inserts.
  • Hygiene Kit to be used with Thunder T3 Ear Muff only
  • Extends performance and lifetime of Ear Muff
  • Recommended replacement every 6 months with general use
  • Snap-in cushions for quick and easy replacement
  • Includes one pair of snap-in ear cushions and one pair of foam ear cup inserts
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Howard Leight™ Ear Muffs
For Use With (Application) For high attenuating noise blocking ear muffs
Product Type Thunder Ear Muff Hygiene Kit
Style Snap-in ear
Includes 2 Replacement ear cushions, 2 Foam inserts