Learn More
Honeywell™ Howard Leight™ Viking™ Ear Muffs
Description
- Fits with other PPE, including hard hats, visors, face shields, respirators and welding shields
- Dielectric construction won't deform
- Snap-in ear cushions
Specifications
Specifications
| Includes | Inner ventilation |
| Certifications/Compliance | ANSI S.3.19-1974 |
| For Use With (Application) | Agriculture and farming, building construction, forestry, general contracting, heavy construction, landscaping, lumber or wood products, metal fabrication Petrochemical Steel Mining Transportation Utility/Waste Management |
| Noise Reduction Rating (NRR) | 29 dB |
| Product Type | Ear Muffs |
| Dielectric | Yes |
| Style | Viking 1 Earmuffs |