Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579388
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579388 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579388 Supplier Invitrogen™ Supplier No. PA579388
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: SW620 whole cell. IHC: Human Pancreatic Cancer Tissue.

In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXA3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene Hoxa3
Gene Accession No. O43365, P02831
Gene Alias homeo box 1E; homeo box A3; homeobox A2; homeobox A3; homeobox protein Hox-1.5; Homeobox protein Hox-1E; homeobox protein Hox-A3; Homeobox protein MO-10; HOX1; Hox-1.5; Hox-1.5-like protein; Hox-15; HOX1E; HOXA3; Hoxa-3; Mo-10; NEWGENE_1561431
Gene Symbols Hoxa3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human HOXA3 (427-443aa HHPSQGRIQEAPKLTHL).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103690130, 15400, 3200
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.