Learn More
Invitrogen™ HOXA6 Monoclonal Antibody (3A6)

Description
Peptide Sequence: GYDALRPFPA SYGASSLPDK TYTSPCFYQQ SNSVLACNRA SYEYGASCFY SDKDLSGASP SGSGKQRGPG DYLHFSPEQQ YKPDSSSGQG KALHDEGADR.
Specifications
Specifications
| Antigen | HOXA6 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 3A6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | HOXA6 |
| Gene Accession No. | P31267 |
| Gene Alias | homeo box 1B; homeo box A6; homeobox A6; homeobox protein Hox-1.2; Homeobox protein Hox-1B; homeobox protein HOXA6; homeobox protein Hox-A6; homeobox protein M5-4; HOX1; HOX1.2; Hox-1.2; HOX1B; Hoxa6; Hoxa-6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.