Learn More
Invitrogen™ HOXB1 Polyclonal Antibody

Description
Immunogen sequence: SAQAVDSYAS EGRYGGGLSS PAFQQNSGYP AQQPPSTLGV PFPSSAPSGY APAACSPSYG PSQYYPLGQS EGDGGYFHPS SYGAQLGGLS DGYGAGGAGP GPYPPQHPPY GNEQTASFAP AYADLLSEDK ETPCPSEPNT PTARTF; Positive Samples: Mouse heart; Cellular Location: Nucleus.
Specifications
Specifications
| Antigen | HOXB1 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.12 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Hoxb1 |
| Gene Accession No. | P14653, P17919 |
| Gene Alias | HCFP3; homeo box B1; homeobox B1; homeobox protein Hox-2.9; Homeobox protein Hox-2I; homeobox protein Hox-B1; HOX2; Hox-2.9; Hox-29; HOX2I; Hoxb1; Hoxb-1 |
| Gene Symbols | Hoxb1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.