Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXB7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 402000
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-200-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-200-0 Supplier Invitrogen™ Supplier No. 402000
  • $500.00
    $375.65 / Each of 1

    Save $124.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

HOXB7 Polyclonal Antibody for Western Blot, ICC/IF

The homeobox genes encode transcription factors that regulate developmental processes such as body patterning and organogenesis. The mammalian HOX gene complex consists of 39 genes that are located on 4 linkage groups (clusters), dispersed over 4 chromosomes (HOXA, HOXB, HOXC and HOXD). HOXB7 is expressed in most malignant T-cell lines and CD4-positive peripheral blood cells, and is constitutively expressed in both melanoma primary lesions and cell lines. HOXB7 binds to and transactivates bFGF in melanoma cell lines but SkBr3 breast adenocarcinoma is negative for the expression of both HOXB7 and bFGF genes. HOXB7 may be a key activator of tumor-associated neoangiogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXB7
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene Hoxb7
Gene Accession No. P09629
Gene Alias AI325018; HHO.C1; homeo box 2C; homeo box B7; homeo box c1 protein; homeobox B7; homeobox gene B7; homeobox protein HHO.C1; Homeobox protein Hox-2.3; homeobox protein Hox-2C; Homeobox protein Hox-B7; homeobox protein MH-22B; Homeobox protein MuB1; homeobox protein R1B; HOX2; Hox-2.3; Hox-23; HOX2C; Hox2r1b; Hoxb7; Hoxb-7; R1b
Gene Symbols Hoxb7
Host Species Rabbit
Immunogen Synthetic peptide derived from the C-terminal region of the human HOXB7 (homeobox B7, homeobox C1) protein.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3217
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.