Learn More
Selleck Chemical LLC (2-Hydroxypropyl)--cyclodextrin (HP--CD) 25mg 128446-35-5 HP--cyclodextrin
SDP
Supplier:  Selleck Chemical LLC S476025MG
Encompass_Preferred
(2-Hydroxypropyl)-?-cyclodextrin (HP-?-CD, HP-?-cyclodextrin), a well-known sugar used in drug delivery, genetic vector, environmental protection and treatment of Niemann-Pick disease type C1 (NPC1), is an inhibitor of amyloid-? aggregation. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8262
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.