Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HP Mouse anti-Human, Clone: UMAB10, liquid, UltraMAB™
SDP

Catalog No. 501685051
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-5051 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-5051 Supplier OriGene Supplier No. UM570010
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Haptoglobin is an a2-globulin plasma protein synthesized in the liver. It is an acute phase protein which aslo acts as a scavenger of hemoglobin from hemolyzed red cells. The haptoglobin monomer consists of two a and two beta chains; the a chanins bay be hp1Fa, hp1Sa, or hp2a types while the beta chain is constant. Three major phenotypes are recognized: HP1-1, Hp2-1 and Hp-2-2, in which the hp1a chain bay be either the F or S type.

Specifications

Antigen HP
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone UMAB10
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene HP
Gene Accession No. P00738
Gene Alias BP, HP2ALPHA2, HPA1S
Gene Symbols HP
Host Species Mouse
Immunogen Full length human recombit protein of human HP produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3240
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.