Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ HP1 alpha Protein
SDP

Catalog No. NBP19886310 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP19886310 100μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP19886310 Supplier Novus Biologicals™ Supplier No. NBP198863100UG
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-191 of Human HP1 alpha The Recombinant Human HP1 alpha Protein is derived from E. coli. The Recombinant Human HP1 alpha Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_001120794
Concentration 1mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 30% glycerol, 0.1M NaCl
Gene ID (Entrez) 23468
Molecular Weight (g/mol) 22kDa
Purification Method SDS-PAGE
Quantity 100μg
Source Recombinant human CBX5 protein, fused to His-tag at N-terminus, expressed in E. coli
Immunogen CBX5, 1-191aa, Human, His tag, E. coli (NP_001120794).
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name HP1 alpha
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.