Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HP1 gamma Monoclonal Antibody (2MOD-1G6)
GREENER_CHOICE

Catalog No. MA3054 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA3054 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA3054 Supplier Invitrogen™ Supplier No. MA3054
  • $648.50
    $502.65 / Each of 1

    Save $145.85

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

MA3-054 detects HP1-gamma in human, mouse and rat samples. MA3-054 has been succesfully used in western blot, ELISA, immunofluorescence and ChIP applications. Immunogen is a HP1-gamma recombinant mouse.

At the nuclear envelope, the nuclear lamina and heterochromatin are adjacent to the inner nuclear membrane. The protein encoded by this gene binds DNA and is a component of heterochromatin. This protein also can bind lamin B receptor, an integral membrane protein found in the inner nuclear membrane. The dual binding functions of the encoded protein may explain the association of heterochromatin with the inner nuclear membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HP1 gamma
Applications ChIP Assay, ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2MOD-1G6
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.05% sodium azide
Gene Cbx3
Gene Accession No. P23198, Q13185
Gene Alias CBX3; chromobox 3; chromobox homolog 3; chromobox homolog 3 (HP1 gamma homolog, Drosophila); chromobox protein homolog 3; chromobox-like protein 3; HECH; heterochromatin protein 1 gamma; heterochromatin protein 1 homolog gamma; heterochromatin protein HP1 gamma; heterochromatin-like protein 1; HGNC:1553; HP1 gamma; HP1 gamma homolog; HP1g; HP1-GAMMA; HP1Hs-gamma; HPy; M32; Modifier 2 protein; OTTHUMP00000202463
Gene Symbols Cbx3
Host Species Mouse
Immunogen Recombinant HP1-gamma mouse.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11335, 12417, 297093
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.