Learn More
Microsolv Technology Corporation HPLC Column, Bidentate C8 2.o, 2.2um, 2.1mm ID x 100mm Length, 120A. 1 EA.
Supplier:  Microsolv Technology Corporation 4020810P2
Encompass_Preferred
This Cogent Bidentate C8 2.o HPLC Column is made with Cogent TYPE-C™ Silica , and this smaller particle, silica hydride based stationary phase provides Increased Efficiency, Excellent Selectivity and Faster Equilibration. Considered "Near UHPLC", this column can be used on any HPLC Instrument and still deliver improved efficiency. Chosen for analysis of hydrophobic compounds that are difficult to retain in Aqueous Mobile Phases, including non-polar compounds when C18 is too retentive. This column is considered a Mass Spec Friendly Column and is one of the more popular 2.o column sizes and often chosen for LCMS when increased resolution is desired.
Catalog No. 50-227-0825
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.