Learn More
Microsolv Technology Corporation Prep HPLC Column, Bidentate C8, 4um, 100A. 30mm ID x 150mm Length. 1 EA.
Supplier:  Microsolv Technology Corporation 40008P30150
Encompass_Preferred
This Cogent Bidentate C8 Preparative, HPLC Column is made with 4um, 100A, Cogent TYPE-C™ Silica , and this silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Very Robust Column Lifetimes. Chosen for Purification or "Prep" of Hydrophobic Molecules, Metabolites, and other Analytes with amine groups when a large volume of sample is desired to be collected and C18 is too retentive. A specialized HPLC System is required for columns this size.
Catalog No. 50-227-0694
Only null left
Add to Cart
Request a Quote