Learn More
MilliporeSigma™ Supelco™ HPLC Column Coupler
Catalog No. 111011349 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
HPLC Column Coupler
Catalog No.
11-101-1349 HPLC Column Coupler
1 options
Catalog No. 11-101-1349 Supplier MilliporeSigma™ Supelco™ Supplier No. 55213
Add to Cart
Add to Cart
Connects two Discovery or SUPELCOSIL analytical columns. Zero dead volume design.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Chromatography Applications