Learn More
Microsolv Technology Corporation HPLC Column, Diamond Hydride, 4um, 3.0mm ID x 50mm Length, 100A. 1 EA.
Supplier:  Microsolv Technology Corporation 7000005P3
Encompass_Preferred
This Cogent Diamond Hydride HPLC Column is made with Cogent TYPE-C™ Silica , and this silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Robust Column Lifetimes. Chosen for analysis of polar compounds that are difficult to retain in Reversed Phase, including polar and non-polar peptides, amino acids and other biological compounds such as metabolites. This size column provides similar run times as a 4.6mm ID column, but uses much less mobile phase, with lower flow rates. No special equipment is needed for this column to save solvent use.
Catalog No. 50-227-0925
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.