Learn More
Microsolv Technology Corporation Prep HPLC Column, UDA (wcx), 4um, 100A. 21.2mm ID x 50mm Length, Preparative. 1 EA.
Supplier:  Microsolv Technology Corporation 40031P2150
Encompass_Preferred
This Cogent UDA™ (wcx) HPLC Column is made with Cogent TYPE-C™ Silica , and has an 11 carbon chain with a terminal carboxylic acid group. This silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Extremely Robust Column Lifetimes.Use in Reversed Phase (RP), Aqueous Normal Phase (ANP) or as a Weak Cation Exchange Column (wcx at pH 6 or higher). Faster equilibration than other Silica columns, no need to control moisture in mobile phase solvents and robust column lifetimes Use Minimal Salt to Elute in ANP A specialized HPLC System is required for columns this size.
Catalog No. 50-227-0748
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.