Learn More
Higgins Analytical Inc HPLC REVERSE PHASE COLUMN, 50X
Supplier:  Higgins Analytical Inc TS05M5C185
Encompass_Preferred
TARGA C18 5um, 50x0.5mm Microbore HPLC Column
Catalog No. NC0389305
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.