Learn More
R&D Systems™ HPSE Substrate

Description
- Used as a substrate for recombinant HPSE activity assay
- Detecting the presence of HPSE and heparinase activity in biological sample
- Ideal for inhibitor screening for HPSE
- Biotinylated at the non-reducing end of heparan sulfate
- The incorporated biotin does not interfere with HPSE activity
- Extremely sensitive to HPSE activity
- Ideal for ELISA testing
- User-friendly
Specifications
Specifications
| Content And Storage | Store the unopened product at ≤ -70 °C. Good for 12 months from date of receipt. Provided in 25 mM Tris, 150 mM NaCl, pH 7.5, 1% BSA |
| Type | Substrate |
| Quantity | 200 μL |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.