Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PSGL-1/CD162 Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. fab9962r100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB9962R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB9962R100 Supplier R&D Systems Supplier No. FAB9962R100UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PSGL-1/CD162 Monoclonal specifically detects PSGL-1/CD162 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen PSGL-1/CD162
Applications Flow Cytometry
Classification Monoclonal
Clone 688124
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q14242
Gene Alias CD162, CD162 antigen, P-selectin glycoprotein ligand 1, PSGL1, PSGL-1cutaneous lymphocyte-associated associated antigen, selectin P ligandCLA, SELPLG
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human PSGL-1/CD162, Gln42-Gly295, Accession # Q14242
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 6404
Test Specificity Detects human PSGL-1/CD162 in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.