Learn More
AdipoGen HPTS
SDP
Supplier:  AdipoGen CDXH0034G005
Encompass_Preferred
Chemical. CAS 6358-69-627928-00-3 free acid. Formula C16H7Na3O10S3. MW 524.39. Synthetic. Useful for optical sensor applications. Fluorescent pH indicator for physiological range.
Catalog No. 50-256-0786
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.