Learn More
Creative Biomart Recombinant HPV-11 Antigen, GST-tagged
Supplier:  Creative Biomart HPV344
Encompass_Preferred
Recombinant HPV-11 antigen is a 58.1 kDa protein covering the full length of HPV-11 major capsid, its N-terminus is fused with a GST tag, having a total molecular weight of 84 kDa. The HPV-11 was puri
Catalog No. 50-208-4722
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.