Learn More
Biocare Medical, LLC HPV Cocktail Broad Spectrum Antibody - Predilute, 6.0 ml
SDP
Supplier:  Biocare Medical, LLC PM177AA
Encompass_Preferred
The broad spectrum HPV antibody was produced (1H8) against SDS-disrupted bovine papillomavirus type 1 (BPV-1) and used to identify the product of the L1 open reading frame (ORF) of BPV-1. IH8 was found to be reactive with purified major capsid protein (MCP). The antibody was tested with ELISA and with an immunofluorescent technique and detected HPV-1, 6, 11, 16, 18, and 31 in formalin-fixed paraffin embedded biopsy specimens. The CAMVIR-1 antibody was raised against the major capsid protein L1 of human papillomavirus type 16, using a recombinant vaccinia virus that expresses the L1 protein, as a target for screening. This antibody reacted with a 56 kilodalton protein in cells infected with L1-vaccinia virus, and the protein was present in HPV16. A panel of p16 and Ki-67 can be used for further evaluation. CAMVIR-1 may also be used separately for phenotyping HPV-16. Other HPV isotypes may also be reactive with the Broad Spectrum HPV antibody, but have not been tested.
Catalog No. 50-830-93
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.