Learn More
Hach Company Hach HQ4200 Portable Multi-Meter with gel pH and conductivity electrodes, 1 m cables
Supplier:  Hach Company LEV015.53.42002
Encompass
The HQ4200 portable multi-parameter meter is a rugged, dual-channel instrument for field and laboratory water quality testing. It supports simultaneous measurement of pH, conductivity, dissolved oxygen, ORP, salinity, TDS, and ion-selective electrodes, and includes data logging, rechargeable battery power, and USB data transfer.
  • Dual probe inputs for simultaneous multi-parameter measurement.
  • Rugged IP67 enclosure for reliable field use.
  • Rechargeable internal battery with USB charging for extended field operations.
  • 1 m probe cables for flexible positioning during sampling.
  • Large color display showing up to three parameters at a time.
  • On-screen guidance for calibration and troubleshooting to simplify procedures.
  • Stores up to 100,000 data points for long-term logging.
  • Includes field accessories and user manuals for out-of-the-box operation.
Catalog No. NC2742856