Learn More
Abcam HQNO, antagonist of dihydrostreptomycin, 10MG
Supplier:  Abcam AB14460410MG
Encompass_Preferred
MW 259.34 Da, Purity >99%. Multifunctional compound. Various pharmacological effects. Respiratory chain inhibitor that blocks mitochondrial complex I, III and NADH oxidase activity. Mitochondrial metabolism modifier. Antagonist of dihydrostreptomycin. Active in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5576
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.