Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HRASLS3 Monoclonal Antibody (OTI1A5)
GREENER_CHOICE

Catalog No. PIMA526290
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526290 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526290 Supplier Invitrogen™ Supplier No. MA526290
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HRASLS3 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

HRASLS3 specifically catalyzes the release of fatty acids from phospholipids in adipose tissue. It also has a weak lysophospholipase activity. Tumor suppressor that may be involved in interferon-dependent cell death.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HRASLS3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI1A5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PLAAT3
Gene Accession No. P53816
Gene Alias Adipose-specific phospholipase A2; adipose-specific PLA2; AdPLA; C78643; Ca-independent phospholipase A1/2; group XVI phospholipase A1/A2; Group XVI phospholipase A2; HRAS like suppressor 3; HRAS-like suppressor 1; HRAS-like suppressor 3; HRASLS3; H-rev 107 protein; H-rev 107 protein homolog; Hrev107; H-REV107; H-REV107-1; HREV107-1; HREV107-3; HRSL3; LRAT-like protein 3; MLP-3; Phospholipase A and acyltransferase 3; phospholipase A2 group XVI; phospholipase A2, group XVI; Pla2g16; PLAAT3; Renal carcinoma antigen NY-REN-65
Gene Symbols PLAAT3
Host Species Mouse
Immunogen Full length human recombinant protein of PLA2G16 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11145
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.