Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HRASLS5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7207442 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-1664 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-1664 Supplier Proteintech Group Inc Supplier No. 175811AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

catalyzes N-acylation of phosphatidylethanolamine (PE) to generate N-Acylphosphatidylethanolamine (NAPE) a precursor of bioactive N-acylethanolamines, including the endocannabinoid adamide.

Specifications

Antigen HRASLS5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HRASLS5
Gene Accession No. Q96KN8, Q9CPX5
Gene Alias H rev107 like protein 5, HRAS like suppressor 5, HRASLS5, HRLP5, iNAT, RLP1
Gene Symbols Hrasls5
Host Species Rabbit
Immunogen HRASLS5 Fusion Protein Ag11113
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117245, 293711, 66727
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.