Learn More
LifeSensors Hrd1 (Synoviolin) 25ug
SDP
Supplier:  LifeSensors UB03070025
Encompass_Preferred
Hrd1 is a RING domain E3 ligase that is involved in the conjugation of ubiquitin to target substrates. Hrd1 has been demonstrated to function with the E2 enzymes UBE2D3 (UbcH5c) and UBE2G2 in vitro. Hrd1 is an ER-associated ligase involved in ERAD and has been linked to rheumatoid arthritis. It is present in human rheumatoid synovial cells and found to be a causative factor for arthropathy in in vivo studies. Hrd1 is also found in brain neurons and its proper regulation may be linked to neurodegenerative diseases. This construct is a Hrd1 N-terminal deletion containing residues 236-617 consisting of the complete C-terminal cytoplasmic portion. The N-terminal transmembrane spanning portion has been deleted. This Hrd1 N-terminal deletion mutant is active as an E3 ligase containing the RING and substrate recognition domains. This product contains 25ug of HRD1 (Synoviolin)
Catalog No. NC2285782
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.