Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ HRP Check&Go! Control Kit
SDP

Catalog No. 40020030 Shop All Novus Products
Change view
Click to view available options
Quantity:
30 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-020-030 30 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-020-030 Supplier R&D Systems™ Supplier No. 40020030

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. HRP Check&Go! is a quick immunochromatography test that confirms the successful conjugation of Horseradish Peroxidase (HRP) to antibodies without the need for any specialized or costly equipment

The key component of the kit is a nitrocellulose membrane containing a Test line of immobilized Protein A and Protein G called a half strip. The HRP-antibody conjugate is run on the Protein A/G Strip. The conjugate binds the Protein A and Protein G concentrated on the Test line. After the addition of the HRP detection solution, a visible line on the strip appears.

Kit Contents:

  • 1 cryovial of HRP detection solution
  • 1 vial of Positive Control
  • 3 cryovials of 10X Running Buffer
  • 30 Protein A/G Strips
  • Not supplied: 96-wells low binding plate, Bovine serum albumin (BSA)

Specifications

Target HRP Check&Go!
Content And Storage Storage of components varies. See protocol for specific instructions.
Product Line Novus
Quantity 30 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.