Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ HRP Conjugate Stabilizer/Diluent
SDP

Catalog No. 9010005 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-100-05 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-100-05 Supplier Novus Biologicals™ Supplier No. 9010005

missing translation for 'validMainframeMsg'

Applications: Western Blot

Specifications

Content And Storage 4°C. Do not freeze.
Quantity 50 mL
Type HRP Conjugate Stabilizer/Diluent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.