Learn More
Bioassay Systems QuantiChrom™ HRP Detection Reagent. A stabilized sensitive reagent for peroxidases, for use in ELISA, immunochromogenic chemistry and Western blot.
Supplier: Bioassay Systems DTMB200
A stabilized sensitive reagent for peroxidases, for use in ELISA, immunochromogenic chemistry and Western blot. Key Features: Highly sensitive. Detects as low as 7.6x10-8 U/well, or 7.7 pg/well horseradish peroxidase (HRP) in 96-well plate assays. Detects as low as 0.1 ppb Gliadin in ELISA. Improved stability and assay robustness. Method: OD450nm. Samples: peroxidases and HRP in ELISA. Species: All. Procedure: Assay takes Approx. 30 min. Kit size: 200 Tests. Detection limit: 76 nU/well, or 7.7 pg/well.
HORSERADISH PEROXIDASE (HRP) is an enzyme commonly used when conjugated to analytes of interest. When combined with hydrogen peroxide and a dye, HRP can be used to colorimetrically quantify the analyte.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.