Learn More
Cytiva HRP labelled Protein A
Catalog No. 45000678 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
45-000-678 1 mL
1 options
Catalog No. 45-000-678 Supplier Cytiva Supplier No. NA91201ML
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
Protein A conjugated with Horseradish peroxidase for detection of immunoglobulins via affinity to the Fc region - typically IgG from many mammalian species
For use in immunodetection and in situ hybridization applications.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Protein A HRP
For Use With (Application) For detection of immunoglobulins via affinity to the Fc region - typically IgG from many mammalian species
Quantity 1 mL