Learn More
AGI NORTH AMERICA INC HS Martin 50L jacketed reactor system installed in a corrosion resistant mobile framework, equipped with 4ft² (.37m²) condenser and electric motor controlled by a Nema 4X VFD
SDP
Supplier:  AGI NORTH AMERICA INC 5003906050F
Encompass_Preferred
Reactor pressure rating: up to 7PSI (.5Bar) to full vacuum-Reactor jacket volume: ~20L-Reactor jacket provides ~7 ft² (.7m²) of heat transfer surface area-Framework constructed with polished SS pipe mounted on lockable, non-marring swivel casters-System rated to handle continuous temperatures of +200 deg C-Glassware is constructed of 33 expansion borosilicate glass-Provided with a cover-mounted thermowell along with 4 unused ports for other accessories-The jacketed reactor has graduation scales on both the front and rear for easy viewing-Flush bottom valve eliminates “dead space” in the bottom of the reactor while providing complete draining of reactor-Wetted materials: either borosilicate glass or PTFE-Oring material: FKM. Other compounds available upon request-Jacket nozzles equipped with M24 threaded adapters to match most circulators-AC 3-phase inverter duty motor, 3/8 HP, 58 in-lbs of torque, variable speed to 340rpm-VFD: NEMA 4X/IP66 inverter w. digital display
Catalog No. NC2417598
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.