Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech Human Serum Albumin (HSA), lyophilized powder, Fatty acid free (50 g)
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0626150
Encompass_Preferred
PAN-Biotech Human Serum Albumin (HSA), Fatty Acid Free lyophilized powder (50 g). APPLICATION: Due to their free hydrophobic region, fatty acid free albumin is used to solubilize lipids in tissue culture. It is also used as a blocking agent in Western blots or ELISA technique and is also an effective cryoprotectant. Fatty acid (FA)-free. FA-free HSA allows for strict control of FAs in cell culture. FA-free HSA maximizes binding sites for the use of specific fatty acids in cell culture. Highly purified product absent of chemical and microbial contamination. Manufactured under conditions certified to ISO 9001. Each donor unit used in the preparation of the product was tested and found negative for Anti-HIV 1/2 and Anti-HCV and nonreactive for HBsAg, HIV-1 Antigen and RPR with kits approved by the FDA. CAS REGISTRY NO.: 70024-90-7. COUNTRY OF ORIGIN: Germany. TYPE: Purified Protein. FATTY ACID CONTENT: Fatty acid free. PROTEASE CONTENT: Not protease free. STORAGE: Store at 2-8°C.
Catalog No. 50-238-8486
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.