Learn More
Medchemexpress LLC hsa-miR-155-5p mimic 20nmol | 20nmol
SDP
Supplier:  Medchemexpress LLC HYR0031620NMOL
Encompass_Preferred
hsa-miR-155-5p mimics are small chemically synthesized double-stranded RNAs that mimic endogenous miRNAs and enable miRNA functional analysis by up-regulation of miRNA activity
Catalog No. 50-003-22443