Learn More
Medchemexpress LLC hsa-miR-17-5p mimic 20nmol | 20nmol
Supplier: Medchemexpress LLC HYR0032620NMOL
hsa-miR-17-5p mimics are small chemically synthesized double-stranded RNAs that mimic endogenous miRNAs and enable miRNA functional analysis by up-regulation of miRNA activity